Novus Biologicals
Manufacturer Code:NBP229731
Catalog # NBP229731
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Radioimmune Assays (RIA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3A11 |
Immunogen | Recombinant fusion protein TrpE-GluR2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: AMPA 2 glutamate receptor ionotropic AMPA 2; AMPA-selective glutamate receptor 2; GluA2; GluR-2; GluR-B; GluR-K2; Glutamate receptor 2; glutamate receptor B flip isoform; Glutamate receptor ionotropic, AMPA 2; glutamate receptor, ionotropic, AMPA 2
Gene Aliases: GluA2; GluR-K2; GLUR2; GLURB; GRIA2; HBGR2
UniProt ID: (Human) P42262
Entrez Gene ID: (Human) 2891
Molecular Function:
ion channel
ionotropic glutamate receptor
ligand-gated ion channel
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.