Invitrogen
Manufacturer Code:MA527694
Catalog # PIMA527694
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S355-1 |
Immunogen | Fusion protein amino acids 1-389 (extracellular N-terminus) of rat GluA1/GluR1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: AMPA 1; AMPA 1 AMPA selective glutamate receptor 1 Glutamate receptor 1 GluA1 GLUH 1 GluR K1 GluRK1 Glr-1 Glur-1 HIPA Glur1 GluR-A AI853806 2900051M01Rik GluRA Glr1 MGC13325 GLUH1 GluA1 GLUR1 HBGR1 GLURA gluR- GluA1 Glutamate receptor ionotropic AMPA 1 Glutamate receptor ionotropic Gria 1 HBGR1 OTTHUMP00000160643 OTTHUMP00000165781 OTTHUMP00000224241 OTTHUMP00000224242 OTTHUMP00000224243; AMPA-selective glutamate receptor 1; GluA1; GluR-1; GluR-A; GluR-K1; GluR1; Glutamate receptor 1; glutamate receptor A; Glutamate receptor ionotropic, AMPA 1; glutamate receptor subunit GluR1; glutamate receptor, ionotropic, AMPA 1; glutamate receptor, ionotropic, AMPA1 (alpha 1)
Gene Aliases: 2900051M01Rik; Glr-1; Glr1; GluA1; GLUH1; Glur-1; GluR-A; gluR-K1; GLUR1; GLURA; GRIA1; HBGR1; HIPA1
UniProt ID: (Human) P42261, (Rat) P19490, (Mouse) P23818
Entrez Gene ID: (Human) 2890, (Rat) 50592, (Mouse) 14799
Molecular Function: ion channel ionotropic glutamate receptor ligand-gated ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.