Invitrogen
Manufacturer Code:MA518117
Catalog # PIMA518117
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | RH95 |
Immunogen | Synthetic peptide corresponding to the N-terminal region of rat GRIA1 conjugated to KLH |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C or -80°C if preferred |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: AMPA-selective glutamate receptor 1; GluA1; GluR-1; GluR-A; GluR-K1; GluR1; Glutamate receptor 1; Glutamate Receptor 1 GRIA1 GluRA HGBR1; glutamate receptor A; Glutamate receptor ionotropic, AMPA 1; glutamate receptor subunit GluR1; glutamate receptor, ionotropic, AMPA1 (alpha 1)
Gene Aliases: 2900051M01Rik; Glr-1; Glr1; GluA1; Glur-1; GluR-A; gluR-K1; Glur1; GluRA; Gria1; HIPA1
UniProt ID: (Rat) P19490, (Mouse) P23818
Entrez Gene ID: (Rat) 50592, (Mouse) 14799
Molecular Function:
ion channel
ionotropic glutamate receptor
ligand-gated ion channel
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.