Invitrogen
Manufacturer Code:PA536405
Catalog # PIPA536405
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 831-880 of human GluR- delta2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: creeper; GluD2; gluR delta-2 subunit; glutamate receptor delta-2 subunit; Glutamate receptor ionotropic delta-2 GluD2 GluR delta-2 subunit GRID2 GLURD2 GluR delta2 GluR delta2; Glutamate receptor ionotropic, delta-2; glutamate receptor, ionotropic, delta 2; hotfoot; lurcher; minisatellite 10ac detected by probe MMS10
Gene Aliases: B230104L07Rik; cpr; GluD2; GLURD2; GluRdelta2; GRID2; ho; Lc; Lc<J>; MMS10-AC; Ms10ac; nmf408; SCAR18; tpr
UniProt ID: (Human) O43424, (Mouse) Q61625, (Rat) Q63226
Entrez Gene ID: (Human) 2895, (Mouse) 14804, (Rat) 79220
Molecular Function:
ion channel
ionotropic glutamate receptor
ligand-gated ion channel
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.