Invitrogen
Manufacturer Code:PA5112429
Catalog # PIPA5112429
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Glutamate dehydrogenase 2 mitochondrial protein (319-558AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GDH 2; GDH 2 GDH2 GLUD2 GLUDP1 GluR delta2 glutamate dehydrogenase 2 Glutamate dehydrogenase 2 mitochondrial glutamate dehydrogenase pseudogene 1 testicular secretory protein Li 14; Glutamate dehydrogenase 2, mitochondrial; glutamate dehydrogenase pseudogene 1; testicular secretory protein Li 14
Gene Aliases: GDH2; GLUD2; GLUDP1
UniProt ID: (Human) P49448
Entrez Gene ID: (Human) 2747
Molecular Function:
dehydrogenase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.