Invitrogen
Manufacturer Code:PA519267
Catalog # PIPA519267
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (ESEEQKRNRVRGILR) corresponding to the internal amino acids of GLUD1 / GLUD2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: epididymis tissue sperm binding protein Li 18mP; GDH 1; GDH 2; GLUD1 glutamate dehydrogenase 1 GDH GDH1 GLUD MGC132003 glutamate dehydrogenase (NAD(P)+) GLUD2 glutamate dehydrogenase 2 GDH2 GLUDP1 glutamate dehydrogenase pseudogene 1; glutamate dehydrogenase (NAD(P)+); Glutamate dehydrogenase 1, mitochondrial; Glutamate dehydrogenase 2, mitochondrial; glutamate dehydrogenase pseudogene 1; Memory-related gene 2 protein; MRG-2; testicular secretory protein Li 14
Gene Aliases: Ac2-281; AI118167; GDH; Gdh-X; GDH1; GDH2; GLUD; GLUD1; GLUD2; Gludeha; Gludl; GLUDP1; MRG-2
UniProt ID: (Human) P00367, (Human) P49448, (Mouse) P26443, (Rat) P10860
Entrez Gene ID: (Human) 2746, (Human) 2747, (Mouse) 14661, (Rat) 24399
Molecular Function: dehydrogenase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.