Invitrogen
Manufacturer Code:MA535668
Catalog # PIMA535668
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC1216 |
Immunogen | A synthesized peptide derived from human GLUD1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: Ac2-281 AI118167 BOS_24262 GDH GDH 1 GDH1 Gdh-X GLUD Glud1 Gludeha Gludl Glutamate dehydrogenase 1 mitochondrial Memory-related gene 2 protein MRG-2; epididymis tissue sperm binding protein Li 18mP; GDH 1; glutamate dehydrogenase (NAD(P)+); Glutamate dehydrogenase 1, mitochondrial; Memory-related gene 2 protein; MRG-2
Gene Aliases: Ac2-281; AI118167; GDH; Gdh-X; GDH1; GLUD; GLUD1; Gludeha; Gludl; MRG-2
UniProt ID: (Human) P00367, (Mouse) P26443, (Rat) P10860
Entrez Gene ID: (Human) 2746, (Mouse) 14661, (Rat) 24399
Molecular Function:
dehydrogenase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.