Invitrogen
Manufacturer Code:PA596899
Catalog # PIPA596899
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 880-1009 of human GRID1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GDH 1; Glutamate dehydrogenase 1, mitochondrial; Memory-related gene 2 protein; MRG-2
Gene Aliases: Ac2-281; AI118167; Gdh-X; Gdh1; Glud; Glud1; Gludeha; Gludl; MRG-2
UniProt ID: (Mouse) P26443, (Rat) P10860
Entrez Gene ID: (Mouse) 14661, (Rat) 24399
Molecular Function:
dehydrogenase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.