Invitrogen
Manufacturer Code:PA569381
Catalog # PIPA569381
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the N terminal region of human Glutamate Dehydrogenase |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: epididymis tissue sperm binding protein Li 18mP; GDH 1; GLUD1 glutamate dehydrogenase 1 GDH GDH1 GLUD MGC132003 glutamate dehydrogenase (NAD(P)+) GLUD2 glutamate dehydrogenase 2 GDH2 GLUDP1 glutamate dehydrogenase pseudogene 1; glutamate dehydrogenase (NAD(P)+); Glutamate dehydrogenase 1, mitochondrial; Membrane protein 50; Memory-related gene 2 protein; MP50; MRG-2
Gene Aliases: Ac2-281; AI118167; GDH; GDH 1; Gdh-X; GDH1; GLUD; GLUD1; Gludeha; Gludl; MP50; MRG-2
UniProt ID: (Human) P00367, (Bovine) P00366, (Pig) P42174, (Mouse) P26443, (Rat) P10860
Entrez Gene ID: (Horse) 100064139, (Human) 2746, (Dog) 100684847, (Rabbit) 100351029, (Bovine) 281785, (Sheep) 443239, (Pig) 100157162, (Mouse) 14661, (Guinea pig) 100722523, (Rat) 24399
Molecular Function: dehydrogenase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.