Abnova
Manufacturer Code:PAB27636
Catalog # 89-129-148
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to internal region of rat Glra1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of Glra1.
For Research Use Only
Protein Aliases: Glycine receptor 48 kDa subunit; Glycine receptor strychnine-binding subunit; Glycine receptor subunit alpha-1; glycine receptor, alpha 1 subunit; GLYRA1
Gene Aliases: Glra1; Glyr; GLYRA1
UniProt ID: (Rat) P07727
Entrez Gene ID: (Rat) 25674
Molecular Function:
GABA receptor
acetylcholine receptor
ion channel
ligand-gated ion channel
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.