Abnova
Manufacturer Code:MAB6768
Catalog # 89-113-032
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 6F10 |
Immunogen | Recombinant GST fusion protein corresponding to full length mouse Glo1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rat monoclonal antibody raised against full-length recombinant Glo1.
For Research Use Only
Protein Aliases: 0610009E22Rik; 1110008E19Rik; 2510049H23Rik; Aldoketomutase; AW550643; Glo-1; Glo-1r; Glo-1s; Glo1-r; Glo1-s; Glx I; GLY1; glyoxalase 1 complex; glyoxalase 1 regulatory; glyoxalase 1 structural; Glyoxalase I; Ketone-aldehyde mutase; Lactoylglutathione lyase; Methylglyoxalase; Qglo; S-D-lactoylglutathione methylglyoxal lyase
Gene Aliases: 0610009E22Rik; 1110008E19Rik; 2510049H23Rik; AW550643; Glo-1; Glo-1r; Glo-1s; Glo1; Glo1-r; Glo1-s; GLY1; Qglo
UniProt ID: (Mouse) Q9CPU0
Entrez Gene ID: (Mouse) 109801
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.