Abnova
Manufacturer Code:PAB0764
Catalog # 89-114-405
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide (conjugated diphtheria toxoid) corresponding to amino acids 253-269 of rat Gja5. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C on dry atmosphere. After reconstitution with deionized water,store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Sheep polyclonal antibody raised against synthetic peptide of Gja5.
For Research Use Only
Protein Aliases: connexin 40; Connexin-40; Cx40; Gap junction alpha-5 protein; gap junction membrane channel protein alpha 5
Gene Aliases: Cx40; Cxn-40; Gja5
UniProt ID: (Rat) P28234
Entrez Gene ID: (Rat) 50563
Molecular Function:
cell junction protein
gap junction
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.