R&D Systems
Manufacturer Code:FAB8370S100UG
Catalog # FAB8370S100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 502430 |
Immunogen | NS0 mouse myeloma cell line transfected with human GHSR Accession # NP_940799 |
Conjugate | Alexa Fluor 750 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Mouse MonoclonalAntibody
For Research Use Only
Protein Aliases: GH-releasing peptide receptor; GH-releasing peptide receptor Ghrelin receptor GHRP GHS-R growth hormone secretagogue receptor growth hormone secretagogue receptor type 1; Ghrelin receptor; GHRP; GHS-R; Growth hormone secretagogue receptor type 1
Gene Aliases: GHDP; GHSR
UniProt ID: (Human) Q92847
Entrez Gene ID: (Human) 2693
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.