R&D Systems
Manufacturer Code:MAB81491100
Catalog # MAB81491100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 226706 |
Immunogen | Synthetic peptide containing human Ghrelin/Obestatin Accession No. Q9UBU3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70°C as supplied. 1 month 2 to 8°C under sterile conditions after reconstitution. 6 months -20 to -70°C under sterile conditions a |
Mouse Monoclonal Antibody
Protein Aliases: Appetite-regulating hormone; appetite-regulating hormone ghrelin ghrelin/obestatin preprohormone ghrelin/obestatin prepropeptide GHRL Growth hormone secretagogue Growth hormone-releasing peptide Motilin-related peptide MTLRPgrowth hormone secretagogue receptor ligand obestatin Protein M46; Ghrelin; ghrelin, growth hormone secretagogue receptor ligand; Ghrelin-27; Ghrelin-28; ghrelin/obestatin preprohormone; Growth hormone secretagogue; Growth hormone-releasing peptide; In2c-preproghrelin; Motilin-related peptide; Obestatin; prepro-appetite regulatory hormone; preproghrelin; Protein M46
Gene Aliases: GHRL; MTLRP; UNQ524/PRO1066
UniProt ID: (Human) Q9UBU3
Entrez Gene ID: (Human) 51738
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.