Novus Biologicals
Manufacturer Code:NB600813
Catalog # NB600813
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-QDILWEEAKEAPADK corresponding to C-Terminus according to NP_057446.1 NP_001128413.1 NP_001128416.1 NP_001128417.1 NP_001128418.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Appetite-regulating hormone; appetite-regulating hormone ghrelin ghrelin/obestatin preprohormone ghrelin/obestatin prepropeptide Growth hormone secretagogue Growth hormone-releasing peptide Motilin-related peptide MTLRPgrowth hormone secretagogue receptor ligand obestatin Protein M46; Ghrelin; ghrelin, growth hormone secretagogue receptor ligand; Ghrelin-27; Ghrelin-28; ghrelin/obestatin preprohormone; Growth hormone secretagogue; Growth hormone-releasing peptide; In2c-preproghrelin; Motilin-related peptide; Obestatin; prepro-appetite regulatory hormone; preproghrelin; Protein M46
Gene Aliases: GHRL; MTLRP; UNQ524/PRO1066
UniProt ID: (Human) Q9UBU3
Entrez Gene ID: (Human) 51738
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.