Invitrogen
Manufacturer Code:PA19597
Catalog # 01-674-471
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Chicken |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse Obestatin peptide (FNAPFDVGIKLSGAQYQQHGRAL) aa 76-98 of Ghrelin precursor. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Chicken Polyclonal Antibody
For Research Use Only
Protein Aliases: Appetite-regulating hormone; Ghrelin; Growth hormone secretagogue; Growth hormone-releasing peptide; Motilin-related peptide; MTLRP obestatin; Obestatin; Obestatin-13; Obestatin-23; preproghrelin; proghrelin; Protein M46
Gene Aliases: 2210006E23Rik; Ghr; Ghrl; m46; MTLRP; MTLRPAP
UniProt ID: (Rat) Q9QYH7, (Mouse) Q9EQX0
Entrez Gene ID: (Rat) 59301, (Mouse) 58991
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.