Invitrogen
Manufacturer Code:PA19596
Catalog # 01-674-470
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Chicken |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 17-28 of human ghrelin. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Chicken Polyclonal Antibody
For Research Use Only
Protein Aliases: Appetite-regulating hormone; Ghrelin; Ghrelin MTLRP; ghrelin, growth hormone secretagogue receptor ligand; Ghrelin-27; Ghrelin-28; ghrelin/obestatin preprohormone; Growth hormone secretagogue; Growth hormone-releasing peptide; In2c-preproghrelin; Motilin-related peptide; Obestatin; prepro-appetite regulatory hormone; preproghrelin; proghrelin; Protein M46
Gene Aliases: 2210006E23Rik; Ghr; GHRL; m46; MTLRP; MTLRPAP; UNQ524/PRO1066
UniProt ID: (Human) Q9UBU3, (Mouse) Q9EQX0
Entrez Gene ID: (Human) 51738, (Mouse) 58991
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.