Novus Biologicals
Manufacturer Code:NBP271556PCP
Catalog # NB271556PCP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3B6 |
Immunogen | Full length recombinant protein of human Gephyrin/GPHN (NP_065857) produced in HEK293T cells. |
Conjugate | PerCP |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: Domain E; Domain G; Gephyrin; gephyrin GPHRYN KIAA1385GPHGEPH; Molybdopterin adenylyltransferase; Molybdopterin molybdenumtransferase; MPT adenylyltransferase; MPT Mo-transferase
Gene Aliases: GEPH; GPH; GPHN; GPHRYN; HKPX1; KIAA1385; MOCODC
UniProt ID: (Human) Q9NQX3
Entrez Gene ID: (Human) 10243
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.