Invitrogen
Manufacturer Code:MA535565
Catalog # PIMA535565
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC1109 |
Immunogen | A synthesized peptide derived from human Gephyrin |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: 5730552E08Rik AI662856 BC027112 C230040D23 Domain E Domain G GEPH Gephyrin GPH GPHN GPHRYN HKPX1 KIAA1385 MOCODC Molybdopterin adenylyltransferase Molybdopterin molybdenumtransferase MPT adenylyltransferase MPT Mo-transferase Putative glycine receptor-tubulin linker protein; Domain E; Domain G; Gephyrin; gephyrin isoform; Molybdopterin adenylyltransferase; Molybdopterin molybdenumtransferase; MPT adenylyltransferase; MPT Mo-transferase; Putative glycine receptor-tubulin linker protein
Gene Aliases: 5730552E08Rik; AI662856; BC027112; C230040D23; GEPH; GPH; GPHN; GPHRYN; HKPX1; KIAA1385; MOCODC
UniProt ID: (Human) Q9NQX3, (Rat) Q03555, (Mouse) Q8BUV3
Entrez Gene ID: (Human) 10243, (Rat) 64845, (Mouse) 268566
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.