Novus Biologicals
Manufacturer Code:NBP104293V
Catalog # NBP104293V
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2H7 |
Immunogen | Recombinant human Geminin (1-209 aa) purified from E. coli |
Conjugate | DyLight 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Gem geminin geminin DNA replication inhibitor RP3-369A17.3; Geminin
Gene Aliases: Gem; GMNN; MGORS6
UniProt ID: (Human) O75496
Entrez Gene ID: (Human) 51053
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.