Invitrogen
Manufacturer Code:MA527752
Catalog # PIMA527752
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | GT1656 |
Immunogen | Recombit protein encompassing a sequence within the center region of human Gelsolin. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Actin-depolymerizing factor; ADF; ADF AGEL GSN Gelsolin; AGEL; Brevin; Gelsolin
Gene Aliases: ADF; AGEL; Gsb; GSN
UniProt ID: (Human) P06396, (Rat) Q68FP1, (Mouse) P13020
Entrez Gene ID: (Human) 2934, (Rat) 296654, (Mouse) 227753
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.