Invitrogen
Manufacturer Code:PA570901
Catalog # PIPA570901
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the N-terminal region of rat Gdpd1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GDPD1 GDE4; Glycerophosphodiester phosphodiesterase 4; Glycerophosphodiester phosphodiesterase domain-containing protein 1; Lysophospholipase D GDPD1
Gene Aliases: 2610020H15Rik; GDE4; GDPD1; RGD1311813
UniProt ID: (Human) Q8N9F7, (Rat) Q0VGK4, (Mouse) Q9CRY7
Entrez Gene ID: (Horse) 100057236, (Pig) 100626512, (Human) 284161, (Dog) 491117, (Rabbit) 100352793, (Bovine) 615890, (Rat) 303407, (Mouse) 66569, (Guinea pig) 100720703
Molecular Function: esterase hydrolase ligase phosphodiesterase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.