Novus Biologicals
Manufacturer Code:NBP297088AF405
Catalog # NB041907
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant Gastric Lipase (AAI12273.1 Met1-Lys398). |
Conjugate | Alexa Fluor 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 3.1.1 EC 3.1.1.3 Gastric lipase gastric triacylglycerol lipase GL HGL HLAL lipase gastric MGC138477 MGC142271; gastric lipase; Gastric triacylglycerol lipase; GL; lipase, gastric
Gene Aliases: GL; HGL; HLAL; LIPF
UniProt ID: (Human) P07098
Entrez Gene ID: (Human) 8513
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.