Novus Biologicals
Manufacturer Code:NBP121724UV
Catalog # NBP121724UV
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | RM0084-6J2 |
Immunogen | Mouse recombinant Gas6 |
Conjugate | DyLight 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Rat Monoclonal Antibody
Protein Aliases: AXL receptor tyrosine kinase ligand; AXL stimulatory factor; AXLLGAXL stimulatory factor AXSFAXL receptor tyrosine kinase ligand DKFZp666G247 FLJ34709 GAS-6 growth arrest-specific 6 growth arrest-specific protein 6; GAS-6; growth arrest-specific 6; Growth arrest-specific protein 6
Gene Aliases: AXLLG; AXSF; GAS6
UniProt ID: (Human) Q14393
Entrez Gene ID: (Human) 2621
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.