R&D Systems
Manufacturer Code:AF885SP
Catalog # AF885SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human Gas6 Asp118-Ala678 Accession # NP_000811 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: AXL receptor tyrosine kinase ligand; AXL stimulatory factor; AXLLG AXLLGAXL stimulatory factor AXSFAXL receptor tyrosine kinase ligand DKFZp666G247 FLJ34709 GAS-6 growth arrest-specific 6 growth arrest-specific protein 6; GAS-6; growth arrest-specific 6; Growth arrest-specific protein 6
Gene Aliases: AXLLG; AXSF; GAS6
UniProt ID: (Human) Q14393
Entrez Gene ID: (Human) 2621
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.