Novus Biologicals
Manufacturer Code:NBP222032IR
Catalog # NBP222032IR
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 11C47 |
Immunogen | This antibody was produced from a hybridoma (rat myeloma fused with spleen cells from a rat immunized with purified recombinant human Gas1. Rat anti human Gas1 NP_002039. |
Conjugate | DyLight 755 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Rat Monoclonal Antibody
Protein Aliases: GAS-1; GAS-1 growth arrest-specific 1 Growth arrest-specific gene-1 growth arrest-specific protein 1; growth arrest-specific 1; Growth arrest-specific gene-1; Growth arrest-specific protein 1
Gene Aliases: GAS1
UniProt ID: (Human) P54826
Entrez Gene ID: (Human) 2619
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.