Abnova
Manufacturer Code:PAB9654
Catalog # 89-134-306
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic phosphopeptide corresponding to residues surrounding S41 of rat Gap43. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic phosphopeptide of Gap43.
For Research Use Only
Protein Aliases: Axonal membrane protein GAP-43; Basp2; brain abundant, membrane attached signal protein 2; growth accentuating protein 43; Growth-associated protein 43; MGC156666; Neuromodulin; Protein F1
Gene Aliases: Basp2; Gap43
UniProt ID: (Rat) P07936
Entrez Gene ID: (Rat) 29423
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.