Proteintech
Manufacturer Code:602341IG150UL
Catalog # 50-173-6335
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2C7F9 |
Immunogen | Gamma cystathionase Fusion Protein Ag2872 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CSE CTH Cystathionine gamma lyase Gamma cystathionase; CTL target antigen; cystathionase (cystathionine gamma-lyase); Cystathionine gamma-lyase; cysteine desulfhydrase; Cysteine-protein sulfhydrase; Gamma-cystathionase; homoserine deaminase; homoserine dehydratase; PRB-RA; Probasin-related antigen
Gene Aliases: 0610010I13Rik; AI314617; BC019483; CSE; CTH; Cys3; H
UniProt ID: (Human) P32929, (Rat) P18757, (Mouse) Q8VCN5
Entrez Gene ID: (Human) 1491, (Rat) 24962, (Mouse) 107869
Molecular Function:
lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.