Novus Biologicals
Manufacturer Code:NBP184319
Catalog # NBP184319
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:HLVLSPPPPHTGPALISALNILEGFNLTSLVSREQALHWVAETLKIALALASRLGDPVYDSTITESMDDMLSKVEAAYLRGHINDSQAAPAPLLPVYELDGAPTAAQVLIMGPDDFIVAMVSSLNQPFGSGLITPSGILLNS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: D20S101 dJ18C9.2 gamma-glutamyltransferase 4 gamma-glutamyltransferase 7 gamma-glutamyltransferase-like 3 gamma-glutamyltransferase-like 5 gamma-glutamyltranspeptidase-like 3 GGT4 GGTL5; gamma-glutamyltransferase 4; Gamma-glutamyltransferase 7; Gamma-glutamyltransferase-like 3; Gamma-glutamyltransferase-like 5; Gamma-glutamyltranspeptidase 7; gamma-glutamyltranspeptidase-like 3; GGT 7; Glutathione hydrolase 7; Glutathione hydrolase 7 heavy chain; Glutathione hydrolase 7 light chain
Gene Aliases: D20S101; dJ18C9.2; GGT4; GGT7; GGTL3; GGTL5
UniProt ID: (Human) Q9UJ14
Entrez Gene ID: (Human) 2686
Molecular Function:
acyltransferase
hydrolase
protease
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.