Novus Biologicals
Manufacturer Code:NBP245619
Catalog # NBP245619
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1D12 |
Immunogen | Full length human recombinant protein of human LGALS9 (NP_002299) produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Ecalectin; ecalectin Gal-9 galectin 9 galectin-9 HUAT lectin galactoside-binding soluble 9 LGALS9A MGC117375 MGC125973 MGC125974 Tumor antigen HOM-HD-21 urate transporter/channel protein; Gal-9; Galectin-9; lectin, galactoside binding soluble 9; lectin, galactoside-binding, soluble, 9; Tumor antigen HOM-HD-21; urate transporter/channel protein
Gene Aliases: HUAT; LGALS9; LGALS9A
UniProt ID: (Human) A7VJG6
Entrez Gene ID: (Human) 3965
Molecular Function: cell adhesion molecule signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.