Novus Biologicals
Manufacturer Code:NBP245618
Catalog # NBP245618
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1C7 |
Immunogen | Full length human recombinant protein of human LGALS3 (NP_002297) produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 35 kDa lectin; advanced glycation end-product receptor 3; Carbohydrate-binding protein 35; Carbohydrate-binding protein 35 CBP 3535 kDa lectin Gal-3 galactin-3 Galactose-specific lectin 3 Galactoside-binding protein GALBPCBP35 galectin-3 GALIG IgE-binding protein L31 L-31 Laminin-binding protein Lectin L-29 lectin galactoside-binding soluble 3 LGALS2 MAC-2 Mac-2 antigen MAC2GAL3; CBP 35; Gal-3; Galactose-specific lectin 3; Galactoside-binding protein; GALBP; Galectin-3; IgE-binding protein; L-31; Laminin-binding protein; Lectin L-29; lectin, galactoside-binding, soluble, 3; MAC-2 antigen
Gene Aliases: CBP35; GAL3; GALBP; GALIG; L31; LGALS2; LGALS3; MAC2
UniProt ID: (Human) P17931
Entrez Gene ID: (Human) 3958
Molecular Function: cell adhesion molecule signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.