Invitrogen
Manufacturer Code:PA579596
Catalog # PIPA579596
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E.coli-derived mouse Galectin 3 recombit protein (Position: K153-I264). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 35 kDa lectin; Carbohydrate-binding protein 35; CBP 35; epsilon BP; Gal-3; Galactose-specific lectin 3; Galectin-3; Galectin-3 Gal-3 35 kDa lectin Carbohydrate-binding protein 35 CBP 35 Galactose-specific lectin 3 IgE-binding protein L-34 galactoside-binding lectin Laminin-binding protein Lectin L-29 Mac-2 antigen Lgals3; IgE binding protein; IgE-binding protein; L-34 galactoside-binding lectin; Laminin-binding protein; lectin galactoside-binding soluble 3 protein; Lectin L-29; lectin, galactose binding, soluble 3; Mac-2 antigen
Gene Aliases: AGE-R3; CBP30; gal-3; gal3; GBP; L-34; Lgals3; Mac-2
UniProt ID: (Rat) P08699
Entrez Gene ID: (Mouse) 16854, (Rat) 83781
Molecular Function: cell adhesion molecule signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.