R&D Systems
Manufacturer Code:MAB1152
Catalog # MAB1152
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 933133 |
Immunogen | <EM>E. coli</EM>-derived recombinant human Galectin-1 Met1-Asp135 Accession # P09382 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 14 kDa laminin-binding protein; 14 kDa laminin-binding protein 14 kDa lectin Beta-galactoside-binding lectin L-14-I BHL DKFZp686E23103 gal-1 GAL1 Galaptin galectin 1 galectin-1 Galectin1 GBP HBL HLBP14 HPL L-14 Lactose-binding lectin 1 Lectin galactoside-binding soluble 1 lectin galactoside-binding soluble 1 LGALS1 Putative MAPK-activating protein PM12 S-Lac lectin 1; 14 kDa lectin; Beta-galactoside-binding lectin L-14-I; beta-galactoside-binding protein 14kDa; Gal-1; Galaptin; Galectin-1; HBL; HLBP14; HPL; Lactose-binding lectin 1; Lectin galactoside-binding soluble 1; lectin, galactoside binding soluble 1; lectin, galactoside-binding, soluble, 1; Putative MAPK-activating protein PM12; S-Lac lectin 1
Gene Aliases: GAL1; GBP; LGALS1
UniProt ID: (Human) P09382
Entrez Gene ID: (Human) 3956
Molecular Function: cell adhesion molecule signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.