Proteintech
Manufacturer Code:602231IG150UL
Catalog # 50-173-6324
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3G10D2 |
Immunogen | Galectin-1 Fusion Protein Ag2432 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 14 kDa laminin binding protein 14 kDa lectin Gal 1 GAL1 Galaptin Galectin 1 Galectin-1 HBL HLBP14 HPL Lactose binding lectin 1 LGALS1 S Lac lectin 1; 14 kDa laminin-binding protein; 14 kDa lectin; Beta-galactoside-binding lectin L-14-I; beta-galactoside-binding protein 14kDa; Gal-1; Galaptin; Galectin-1; HBL; HLBP14; HPL; Lactose-binding lectin 1; Lectin galactoside-binding soluble 1; lectin, galactoside binding soluble 1; lectin, galactoside-binding, soluble, 1; Putative MAPK-activating protein PM12; S-Lac lectin 1
Gene Aliases: GAL1; GBP; LGALS1
UniProt ID: (Human) P09382
Entrez Gene ID: (Human) 3956
Molecular Function: cell adhesion molecule signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.