Invitrogen
Manufacturer Code:PA527455
Catalog # PIPA527455
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 135 of Human Galectin 1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 14 kDa laminin-binding protein; 14 kDa lectin; beta-galactoside binding protein; Beta-galactoside-binding lectin L-14-I; beta-galactoside-binding protein 14kDa; Gal-1; GAL1 GBP 14 kDa laminin-binding protein 14 kDa lectin HBL HLBP14 HPL S-Lac lectin 1 beta-galactoside-binding lectin L-14-I beta-galactoside-binding protein 14kDa gal-1 galaptin galectin 1 galectin-1 lactose-binding lectin 1 putative MAPK-activating protein PM12; Galaptin; Galectin-1; HBL; HLBP14; HPL; Lactose-binding lectin 1; Lectin galactoside-binding soluble 1; lectin, galactoside binding soluble 1; lectin, galactoside-binding, soluble, 1; Putative MAPK-activating protein PM12; S-Lac lectin 1
Gene Aliases: AA410090; Gal-1; GAL1; Galbp; galectin-1; GBP; L-14.5; L14; Lect14; LGALS1
UniProt ID: (Human) P09382, (Mouse) P16045
Entrez Gene ID: (Human) 3956, (Mouse) 16852
Molecular Function: cell adhesion molecule signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.