Novus Biologicals
Manufacturer Code:NB1002846
Catalog # NB1002846
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-QGPEPREGPVHGGE corresponding to C-Terminus according to NP_003605.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: GAL3-R; GAL3-R galanin receptor 3 galanin receptor type 3 galanin receptor family member 3 GALNR3 GALR-3; Galanin receptor type 3; galanin receptor, family member 3; GALR-3
Gene Aliases: GALNR3; GALR3
UniProt ID: (Human) O60755
Entrez Gene ID: (Human) 8484
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.