Novus Biologicals
Manufacturer Code:NB10057079
Catalog # NB10057079
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-HLSDTKENKSRID corresponding to C-Terminus according to NP_001471.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: GAL1-R; galanin receptor 1 galanin receptor type 1 GALNRGALNR1GAL1-R GALR-1; Galanin receptor type 1; GALR-1
Gene Aliases: GALNR; GALNR1; GALR1
UniProt ID: (Human) P47211
Entrez Gene ID: (Human) 2587
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.