R&D Systems
Manufacturer Code:MAB5854SP
Catalog # MAB5854SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 581403 |
Immunogen | <em>E.coli</em>-derived recombinant human Galanin Ala20-Ser123 Accession # P22466 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: GAL GAL1 galanin galanin prepropeptide GALN GALNgalanin-related peptide GLNNgalanin-message-associated peptide GMAP MGC40167; Galanin; Galanin message-associated peptide; Galanin peptides; galanin prepropeptide; galanin-message-associated peptide; galanin-related peptide; galanin/GMAP prepropeptide; GMAP
Gene Aliases: ETL8; GAL; GAL-GMAP; GAL1; GALN; GLNN; GMAP
UniProt ID: (Human) P22466
Entrez Gene ID: (Human) 51083
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.