Abnova
Manufacturer Code:MAB8567
Catalog # 89-113-498
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S87-25 |
Immunogen | Recombinant protein corresponding to amino acids 370-433 of mouse Gabrb3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against partial native Gabrb3.
For Research Use Only
Protein Aliases: A230092K12Rik; AW049585; beta3; cleft palate 1; Cp1; GABA(A) receptor subunit beta-3; Gabrb-3; gamma-aminobutyric acid (GABA-A) receptor, subunit beta 3; Gamma-aminobutyric acid receptor subunit beta-3
Gene Aliases: A230092K12Rik; AW049585; beta3; Cp1; Gabrb-3; Gabrb3
UniProt ID: (Mouse) P63080
Entrez Gene ID: (Mouse) 14402
Molecular Function: GABA receptor acetylcholine receptor ion channel ligand-gated ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.