Abnova
Manufacturer Code:PAB9686
Catalog # 89-134-334
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to rat Gabrb2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against a recombinant Gabrb2.
For Research Use Only
Protein Aliases: GABA(A) receptor subunit beta-2; gamma-aminobutyric acid (GABA-A) receptor, subunit beta 2; Gamma-aminobutyric acid receptor beta 2; Gamma-aminobutyric acid receptor subunit beta-2; gamma-aminobutyric acid receptor, subunit beta 2; GARB2
Gene Aliases: Gabrb-2; Gabrb2; GARB2
UniProt ID: (Rat) P63138
Entrez Gene ID: (Rat) 25451
Molecular Function: GABA receptor acetylcholine receptor ion channel ligand-gated ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.