Abnova
Manufacturer Code:MAB18279
Catalog # 89-338-204
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S93A-49 |
Immunogen | Recombinant protein corresponding to amino acids 873-977 at C-terminus of rat Gabbr1. |
Conjugate | ATTO594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: -; GABA-B receptor 1; GABA-B-R1; GABA-BR1; GABABR1; Gamma-aminobutyric acid type B receptor subunit 1; gb1
Gene Aliases: Gabbr1
UniProt ID: (Rat) Q9Z0U4
Entrez Gene ID: (Rat) 81657
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.