Invitrogen
Manufacturer Code:PA542763
Catalog # PIPA542763
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human GYS2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Glycogen [starch] synthase, liver; Glycogen synthase 2 (liver); glycogen synthase, liver
Gene Aliases: BC021322; cb765; GLYSN; GYS2; LGS; zgc:112057
UniProt ID: (Human) P54840, (Mouse) Q8VCB3
Entrez Gene ID: (Pig) 100157080, (Human) 2998, (Horse) 100064264, (Bovine) 537451, (Rabbit) 100352840, (Dog) 403737, (Mouse) 232493, (Guinea pig) 100720245, (Rat) 25623, (Zebrafish) 373082
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.