Invitrogen
Manufacturer Code:PA5115297
Catalog # PIPA5115297
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human Glycogen synthase 2 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BC021322 Glycogen [starch] synthase liver glycogen liver glycogen synthase 2 glycogen synthase 2 (liver) glycogen synthase liver GLYSN GYS2 LGS; Glycogen [starch] synthase, liver; glycogen synthase 2 (liver)
Gene Aliases: GYS2
UniProt ID: (Human) P54840
Entrez Gene ID: (Human) 2998
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.