Invitrogen
Manufacturer Code:MA515022
Catalog # PIMA515022
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | J.18.5 |
Immunogen | Synthetic peptide corresponding to the sequence of human glycogen synthase |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: glycogen &lsqbstarch] synthase muscle; Glycogen [starch] synthase, muscle; glycogen synthase 1 (muscle); glycogen synthase 3, brain
Gene Aliases: GSY; GYS; GYS1; Gys3; MGS
UniProt ID: (Human) P13807, (Rat) A2RRU1, (Mouse) Q9Z1E4
Entrez Gene ID: (Human) 2997, (Rat) 690987, (Mouse) 14936
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.