Abnova
Manufacturer Code:MAB10677
Catalog # 89-108-876
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3A7 |
Immunogen | Recombinant protein corresponding to human GYS1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against partial recombinant GYS1.
For Research Use Only
Protein Aliases: Glycogen [starch] synthase, muscle; glycogen synthase 1 (muscle); GSY; GYS
Gene Aliases: GSY; GYS; GYS1
UniProt ID: (Human) P13807
Entrez Gene ID: (Human) 2997
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.