Invitrogen
Manufacturer Code:PA5116508
Catalog # PIPA5116508
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 234-333 of human GYG1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AU017667 glycogenin glycogenin 1 glycogenin glucosyltransferase Glycogenin-1 GN1 GN-1 GSD15 Gyg Gyg1; glycogenin 1; glycogenin glucosyltransferase; Glycogenin-1; GN-1; GN1
Gene Aliases: AU017667; GSD15; GYG; GYG1
UniProt ID: (Human) P46976, (Mouse) Q9R062, (Rat) O08730
Entrez Gene ID: (Human) 2992, (Mouse) 27357, (Rat) 81675
Molecular Function: glycosyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.