Invitrogen
Manufacturer Code:PA571572
Catalog # PIPA571572
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 411-440 amino acids from the C-terminal region of human GLT8D3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Glucoside xylosyltransferase 1; Glucoside xylosyltransferase 1 242n2 Glycosyltransferase 8 domain-containing protein 3 GXYLT1 GLT8D3; glycosyltransferase 8 domain containing 3; Glycosyltransferase 8 domain-containing protein 3
Gene Aliases: GLT8D3; GXYLT1
UniProt ID: (Human) Q4G148
Entrez Gene ID: (Human) 283464
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.