Invitrogen
Manufacturer Code:PA570903
Catalog # PIPA570903
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the C-terminal region of human GXYLT1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Glucoside xylosyltransferase 1; glycosyltransferase 8 domain containing 3; Glycosyltransferase 8 domain-containing protein 3; Glycosyltransferase 8 domain-containing protein 3-like protein; GXYLT1 GLT8D3; S33-D
Gene Aliases: GLT8D3; Gm1228; Gm87; GXYLT1
UniProt ID: (Human) Q4G148, (Mouse) Q3UHH8, (Rat) Q6GX83
Entrez Gene ID: (Human) 283464, (Dog) 486604, (Horse) 100054009, (Bovine) 617596, (Rabbit) 100341344, (Mouse) 223827, (Rat) 300173, (Guinea pig) 100716393
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.