Invitrogen
Manufacturer Code:PA554240
Catalog # PIPA554240
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human GULP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CED 6; CED-6 GULP1 CED-6 protein GULP engulfment adaptor PTB domain containing 1 CED6 FLJ31156 OTTHUMP00000205908 OTTHUMP00000205909 OTTHUMP00000205911 PTB domain adaptor protein CED-6 engulfment adapter protein; Cell death protein 6 homolog; engulfment adapter protein; Protein GULP; PTB domain adapter protein CED-6; PTB domain adaptor protein CED-6; PTB domain-containing engulfment adapter protein 1
Gene Aliases: 3110030A04Rik; 5730529O06Rik; CED-6; CED6; GULP; Gulp-2; GULP1
UniProt ID: (Human) Q9UBP9, (Mouse) Q8K2A1, (Rat) Q5PQS4
Entrez Gene ID: (Human) 51454, (Mouse) 70676, (Rat) 314543
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.