Invitrogen
Manufacturer Code:PA521865
Catalog # PIPA521865
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to a region within amino acids 241 and 304 of GULP1 (Uniprot IDNo.Q9UBP9) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CED 6; CED-6 CED6 GULP PTB domain adapter protein CED-6 PTB domain adaptor protein CED-6 PTB domain-containing engulfment adapter protein 1 cell death protein 6 homolog engulfment adapter protein; Cell death protein 6 homolog; engulfment adapter protein; Protein GULP; PTB domain adapter protein CED-6; PTB domain adaptor protein CED-6; PTB domain-containing engulfment adapter protein 1
Gene Aliases: 3110030A04Rik; 5730529O06Rik; CED-6; CED6; GULP; Gulp-2; GULP1
UniProt ID: (Human) Q9UBP9, (Mouse) Q8K2A1, (Rat) Q5PQS4
Entrez Gene ID: (Human) 51454, (Mouse) 70676, (Rat) 314543
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.